Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663740_s1
          See other TAS2R139 GE Assays ›
          Gene Symbol:
          Tas2r139
          Gene Name:
          taste receptor, type 2, member 139
          Gene Aliases:
          Tas2r39, mt2r34
          Chromosome Location:
          Chr.6: 42140936 - 42141895 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663740_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r139
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181275.1 NP_851792.1 1 846 122

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r139

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060657 NM_181275.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84195 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123482 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060657 NM_181275.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P254141 NM_181275.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123482 - -
          Whole Mouse Genome G4122A A_52_P254141 NM_181275.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105229 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123482 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05720305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05720315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05711347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05720298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05720316 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05720306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05236911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05587254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537709 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537707 - -
          Affymetrix Mouse Clariom S Array PSR0600003598.mm.1 NM_181275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003598.mm.1 NM_181275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003598.mm.1 TC0600000452.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 433398 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920463 - -
          Illumina mouse-6 v1.0 expression beadchip 4920463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221481 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221481 - -
          mouse-6 v1.1 (Illumina) scl1733.1.1_289-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1733.1.1_289-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221481 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221481 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline