Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00678891_m1
          See other SEC22A GE Assays ›
          Gene Symbol:
          Sec22a
          Gene Name:
          SEC22 vesicle trafficking protein homolog A (S. cerevisiae)
          Gene Aliases:
          1810005C06Rik, Sec22l2
          Chromosome Location:
          Chr.16: 35311131 - 35363934 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00678891_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sec22a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133704.4 NP_598465.2 3 - 4 462 126
          GenBank mRNA AK007341.1 - 3 - 4 402 126
          AK044615.1 - 3 - 4 397 126
          AK082516.1 - 3 - 4 462 126
          BC010189.1 - 3 - 4 191 126
          BC138969.1 - 2 - 3 376 8861991 126
          BC138971.1 - 2 - 3 376 8861993 126
          BC145477.1 - 2 - 3 378 126
          BC145478.1 - 2 - 3 378 9054040 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001545.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sec22a

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ER to Golgi vesicle-mediated transport
          protein transport
          vesicle-mediated transport
          vesicle fusion with Golgi apparatus
          SNARE binding
          SNAP receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600012684.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012684.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012684.mm.1 TC1600001545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012685.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012685.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012685.mm.1 TC1600001545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012690.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012690.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012690.mm.1 TC1600001545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012686.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012686.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012686.mm.1 TC1600001545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012683.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012683.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012683.mm.1 TC1600001545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012697.mm.1 NM_133704.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012697.mm.1 NM_133704.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012697.mm.1 TC1600001545.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline