Search
Search

Rab26| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177375.1 | NP_796349.1 | 4 - 5 | 587 | 78 | |
| GenBank mRNA | AB232621.2 | - | 4 - 5 | 423 | 78 | |
| AK043912.1 | - | 4 - 5 | 552 | 78 | ||
| BC094931.1 | - | 4 - 5 | 495 | 6436242 | 78 | |
| BC118018.1 | - | 4 - 5 | 728 | 40103722 | 78 | |
Set Membership: |
|