| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NM_001159609.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NM_001159609.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NM_001159610.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NM_001159610.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NR_027510.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
NR_027510.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037581.mm.1 |
TC0200004465.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159609.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159609.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159610.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159610.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_001159612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_025657.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NM_025657.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NR_027510.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
NR_027510.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037567.mm.1 |
TC0200004465.mm.1 |
MTA TC |