Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00713127_m1
          See other AF251705 GE Assays ›
          Gene Symbol:
          AF251705
          Gene Name:
          cDNA sequence AF251705
          Gene Aliases:
          CLM4, Cd300c, Cd300d, DIgR1, Igsf7, Lmir2, MAIR-II
          Chromosome Location:
          Chr.11: 114996721 - 115004357 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00713127_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AF251705
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134158.1 NP_598919.1 1 - 2 392 94
          GenBank mRNA AB091767.1 - 1 - 2 123 94
          AB091768.1 - 1 - 2 106 94
          AK050008.1 - 1 - 2 120 94
          AK154808.1 - 1 - 2 112 94
          AY048685.1 - 1 - 2 392 94
          AY457050.1 - 1 - 2 59 94
          BC006801.1 - 1 - 2 123 3152857 94
          BC111820.1 - 1 - 2 58 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004054.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AF251705

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          positive regulation of cytokine secretion
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978216 AK079395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978216 AY048685.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978216 BC006801.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978216 AF251705.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978216 AB091767.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777298 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157556 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978216 AK079395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978216 AY048685.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978216 BC006801.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978216 AF251705.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978216 AB091767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484020 AK079395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484020 AY048685.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484020 BC006801.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484020 AF251705.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484020 AB091767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484027 AK079395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484027 AK154808.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484027 AY048685.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484027 AY457050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484027 BC006801.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28150821 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157556 - -
          Whole Mouse Genome G4122A A_51_P484020 AK079395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484020 AY048685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484020 BC006801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484020 AF251705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484020 AB091767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484027 AK079395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484027 AK154808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484027 AY048685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484027 AY457050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484027 BC006801.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118679 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157556 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09213874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09213876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09423473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09213870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09423472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28150763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09211855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09213871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09389425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09733503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09149438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09733504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09389420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09733497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09389428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28150786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09213873 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10392818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392820 - -
          Affymetrix HT MG-430 PM Array Plate 1428018_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10392815 - -
          Mouse Genome 430 2.0 Array 1428018_a_at - -
          Mouse Genome 430 2.0 Array 1428018_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140497_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140497_at - -
          Affymetrix Mouse Clariom S Array PSR1100038034.mm.1 NM_134158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038034.mm.1 NM_134158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038034.mm.1 TC1100004054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038037.mm.1 NM_134158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038037.mm.1 NM_134158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038037.mm.1 TC1100004054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038036.mm.1 NM_134158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038036.mm.1 NM_134158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038036.mm.1 TC1100004054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038032.mm.1 NM_134158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038032.mm.1 NM_134158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038032.mm.1 TC1100004054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038033.mm.1 NM_134158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038033.mm.1 NM_134158.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038033.mm.1 TC1100004054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038031.mm.1 TC1100004054.mm.1 MTA TC
          Mouse Expression Set 430 1428018_a_at - -
          Mouse Expression Set 430 1428018_PM_a_at - -
          Murine Genome U74v2 Set 104023_at - -
          Murine Genome U74v2 Set 104023_PM_at - -
          Mouse Genome 430A 2.0 Array 1428018_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428018_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428018_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428018_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 441728 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2647336 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2647336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline