Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00723466_g1
          See other NRN1L GE Assays ›
          Gene Symbol:
          Nrn1l
          Gene Name:
          neuritin 1-like
          Gene Aliases:
          Cpg15-2, G630049C14Rik
          Chromosome Location:
          Chr.8: 105893567 - 105895023 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00723466_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrn1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301174.1 NP_001288103.1 2 - 3 213 55
          NM_175024.4 NP_778189.1 2 - 3 216 55
          GenBank mRNA AK080711.1 - 2 - 3 211 55
          AK090312.1 - 2 - 3 214 55
          BC100315.1 - 2 - 3 216 30931461 55
          CK030777.1 - 2 - 3 197 55
          DQ176852.1 - 2 - 3 203 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001232.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrn1l

          Gene Ontology Categories:

          Function(s)

          nervous system development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096942 BC100315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096942 AK080711.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92466 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101102 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096942 BC100315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096942 AK080711.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136595 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101102 - -
          Whole Mouse Genome G4122A A_52_P86892 NM_175024.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86892 AK080711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86892 AK090312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86892 BC100315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86892 DQ176852.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101022 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07297804 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07297794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07297804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574938 - -
          Affymetrix HT MG-430 PM Array Plate 1456819_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574934 - -
          Mouse Genome 430 2.0 Array 1456819_PM_at - -
          Mouse Genome 430 2.0 Array 1456819_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234700_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234700_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800009874.mm.1 NM_175024.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009874.mm.1 TC0800001232.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009876.mm.1 NM_175024.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009876.mm.1 TC0800001232.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009875.mm.1 NM_175024.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009875.mm.1 TC0800001232.mm.1 MTA TC
          Mouse Expression Set 430 1456819_PM_at - -
          Mouse Expression Set 430 1456819_at - -
          Affymetrix HT Mouse Genome 430B Array 1456819_at - -
          Affymetrix HT Mouse Genome 430B Array 1456819_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617212 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6660372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655905 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655905 - -
          mouse-6 v1.1 (Illumina) scl33399.3.65_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 6660372 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33399.3.65_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655905 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655905 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline