Search
Search

Prr11| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175563.5 | NP_780772.1 | 8 - 9 | 1018 | 90 | |
| GenBank mRNA | AK040348.1 | - | 8 - 9 | 996 | 90 | |
| AK047463.1 | - | 8 - 9 | 996 | 90 | ||
| AK083651.1 | - | 8 - 9 | 1013 | 90 | ||
| BC023918.1 | - | 8 - 9 | 1028 | 5323297 | 90 | |
Set Membership: |
|