Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724127_m1
          See other D11WSU47E GE Assays ›
          Gene Symbol:
          D11Wsu47e
          Gene Name:
          DNA segment, Chr 11, Wayne State University 47, expressed
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 113684400 - 113727444 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724127_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D11Wsu47e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177777.5 NP_808445.2 2 - 3 1848 74
          GenBank mRNA AK017508.1 - 2 - 3 1714 74
          AK139389.1 - 3 - 4 1804 74
          AK171307.1 - 2 - 3 1757 74
          BC026908.1 - 1 - 2 1461 3988907 74
          BC048942.1 - 2 - 3 1851 6442008 74
          BC060257.1 - 1 - 2 717 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001863.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene D11Wsu47e

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143431 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134878 BC048942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134878 BC026908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134878 BC060257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134878 AK171307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134878 NM_177777.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050857 BC048942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050857 BC026908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050857 BC060257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050857 AK139389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050857 AK171307.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132318 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173151 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134878 BC048942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134878 BC026908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134878 BC060257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134878 AK171307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134878 NM_177777.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050857 BC048942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050857 BC026908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050857 BC060257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050857 AK139389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050857 AK171307.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28143441 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07498088 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02433946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07498003 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07498019 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07498080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173151 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132318 - -
          Whole Mouse Genome G4122A A_52_P37634 BC048942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37634 BC026908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37634 BC060257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37634 AK139389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37634 AK171307.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134878 BC048942.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134878 BC026908.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134878 BC060257.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134878 AK171307.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134878 NM_177777.5 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173151 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132318 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09700052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09441004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28143432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28143542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09599779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09599778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09717157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09599775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09700053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09599773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09221393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09700052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28143471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382363 - -
          Affymetrix HT MG-430 PM Array Plate 1452805_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382360 - -
          Mouse Genome 430 2.0 Array 1452805_PM_at - -
          Mouse Genome 430 2.0 Array 1452805_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276852_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276852_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100017456.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017456.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017456.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017461.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017461.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017461.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017457.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017457.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017457.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017460.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017460.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017460.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017462.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017462.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017462.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017464.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017464.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017464.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017454.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017454.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017454.mm.1 TC1100001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017463.mm.1 NM_177777.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017463.mm.1 NM_177777.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017463.mm.1 TC1100001863.mm.1 MTA TC
          Mouse Expression Set 430 1452805_at - -
          Mouse Expression Set 430 1452805_PM_at - -
          Murine Genome U74v2 Set 169955_PM_at - -
          Murine Genome U74v2 Set 168794_at - -
          Murine Genome U74v2 Set 168794_PM_at - -
          Murine Genome U74v2 Set 169955_at - -
          Affymetrix HT Mouse Genome 430B Array 1452805_at - -
          Affymetrix HT Mouse Genome 430B Array 1452805_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 313470 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1170215 - -
          Illumina mouse-6 v1.0 expression beadchip 1500347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700833 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230030 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230030 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700833 - -
          mouse-6 v1.1 (Illumina) scl0001420.1_0-S - -
          mouse-6 v1.1 (Illumina) scl40755.5.1_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170215 - -
          Illumina Sentrix mouse-6.v1 bead array 1500347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40755.5.1_55-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001420.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170215 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230030 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2700833 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230030 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2700833 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40755.5.1_55-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline