Search
Search

Klhl2| Gene Symbol: Klhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178633.3 | NP_848748.2 | 1736 | 102 | ||
| GenBank mRNA | AK020050.1 | - | 387 | 102 | ||
| AK032116.1 | - | 1284 | 102 | |||
| AK167903.1 | - | 1527 | 102 | |||
| AK220570.1 | - | 1736 | 102 | |||
| BC024572.1 | - | 903 | 102 | |||
| BC031142.1 | - | 1607 | 4972965 | 102 | ||
| BC031144.1 | - | 1630 | 4973325 | 102 | ||
Set Membership: |
|