Search
Search

Inpp5f| Gene Symbol: Inpp5f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178641.5 | NP_848756.2 | 13 - 14 | 1800 | 73 | |
| GenBank mRNA | AK030870.1 | - | 13 - 14 | 1802 | 73 | |
| AK129249.1 | - | 13 - 14 | 1757 | 73 | ||
| AK217446.1 | - | 2 - 3 | 173 | 73 | ||
| BC050127.1 | - | 1 - 2 | 161 | 73 | ||
| BC125437.1 | - | 13 - 14 | 1688 | 40129952 | 73 | |
| BC137700.1 | - | 13 - 14 | 1688 | 8860719 | 73 | |
Set Membership: |
|