Search
Search

Clca2| Gene Symbol: Clca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178697.4 | NP_848812.1 | 12 - 13 | 2221 | 88 | |
| GenBank mRNA | AK028704.1 | - | 12 - 13 | 2222 | 88 | |
| AK048276.1 | - | 12 - 13 | 2212 | 88 | ||
| AY161007.1 | - | 12 - 13 | 2177 | 88 | ||
| BC096379.1 | - | 12 - 13 | 2284 | 5317084 | 88 | |
Set Membership: |
|