Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00725448_s1
          See other RPLP0 GE Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          36B4, Arbp, L10E
          Chromosome Location:
          Chr.5: 115559467 - 115563729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00725448_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007475.5 NP_031501.1 7 1226 124
          GenBank mRNA AK010267.1 - 7 1179 124
          BF021407.1 - 1 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp0

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosome biogenesis
          cellular response to interleukin-4
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010221 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135098 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970866 BC082775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105288 BF021407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105288 AK010267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105288 AK009767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105288 NM_007475.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847729 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135098 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970866 BC082775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105288 BF021407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105288 AK010267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105288 AK009767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105288 NM_007475.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134602 BC028444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134602 BC087887.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134602 BC089496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134602 BC057295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P134602 BC099384.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P586574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24010223 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05706693 - -
          Whole Mouse Genome G4122A A_52_P586574 - -
          Whole Mouse Genome G4122A A_51_P134602 BC028444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134602 BC087887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134602 BC089496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134602 BC057295.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134602 BC099384.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131988 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100653 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105288 BF021407.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105288 AK010267.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105288 AK009767.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105288 NM_007475.5 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153224 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04530277 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04530278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04936351 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24010226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24010280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04519415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04530273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04209861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04530271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04430095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24010208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04362185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581029 - -
          Affymetrix HT MG-430 PM Array Plate 1419441_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581027 - -
          Affymetrix Mouse Gene 1.1 ST Array 10524718 - -
          Affymetrix Mouse Gene 1.1 ST Array 10436694 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405935 - -
          Mouse Genome 430 2.0 Array 1419441_PM_at - -
          Mouse Genome 430 2.0 Array 1419441_at - -
          Affymetrix Mouse Clariom S Array PSR0500010962.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010962.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010962.mm.1 NM_007475.5 Refseq
          Mouse Expression Set 430 1419441_at - -
          Mouse Expression Set 430 1419441_PM_at - -
          Murine Genome U74v2 Set 101213_at - -
          Murine Genome U74v2 Set 101213_PM_at - -
          Murine Genome U74v2 Set 170488_PM_r_at - -
          Murine Genome U74v2 Set 170488_r_at - -
          Mouse Genome 430A 2.0 Array 1419441_PM_at - -
          Mouse Genome 430A 2.0 Array 1419441_at - -
          Affymetrix HT Mouse Genome 430A Array 1419441_at - -
          Affymetrix HT Mouse Genome 430A Array 1419441_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 451704 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5390152 - -
          MouseWG-6 v2 Expression BeadChip 5550373 - -
          Illumina mouse-6 v1.0 expression beadchip 5390152 - -
          Illumina mouse-6 v1.0 expression beadchip 5550373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744356 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744356 - -
          mouse-6 v1.1 (Illumina) scl27355.8.1_168-S - -
          mouse-6 v1.1 (Illumina) scl0011837.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550373 - -
          Illumina Sentrix mouse-6.v1 bead array 5390152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011837.1_330-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27355.8.1_168-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744356 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249815 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249815 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744356 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0011837.1_330-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline