Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00726840_s1
          See other SPRR2J-PS GE Assays ›
          Gene Symbol:
          Sprr2j-ps
          Gene Name:
          small proline-rich protein 2J, pseudogene
          Gene Aliases:
          Sprr2j
          Chromosome Location:
          Chr.3: 92418087 - 92419396 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00726840_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sprr2j-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003185.1 2 541 90
          GenBank mRNA AK019074.1 - 2 543 90
          AY158994.1 - 2 546 90
          BC125542.1 - 2 517 90
          BC125544.1 - 2 517 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125451 NR_003185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125451 AK019074.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244212 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113559 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169929 - -
          Whole Mouse Genome G4122A A_51_P491758 BC125544.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491758 NR_003185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491758 AK019074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491758 AY158994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491758 BC125542.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02301868 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493883 - -
          Affymetrix HT MG-430 PM Array Plate 1450811_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493882 - -
          Mouse Genome 430 2.0 Array 1450811_PM_at - -
          Mouse Genome 430 2.0 Array 1450811_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20764_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20764_at - -
          Mouse Expression Set 430 1450811_PM_at - -
          Mouse Expression Set 430 1450811_at - -
          Murine Genome U74v2 Set 101755_PM_f_at - -
          Murine Genome U74v2 Set 101755_f_at - -
          Mouse Genome 430A 2.0 Array 1450811_PM_at - -
          Mouse Genome 430A 2.0 Array 1450811_at - -
          Affymetrix HT Mouse Genome 430A Array 1450811_at - -
          Affymetrix HT Mouse Genome 430A Array 1450811_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 624835 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2749198 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2749198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline