Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00727241_s1
          See other ALYREF2 GE Assays ›
          Gene Symbol:
          Alyref2
          Gene Name:
          Aly/REF export factor 2
          Gene Aliases:
          C130042O11Rik, REF2, Refbp2
          Chromosome Location:
          Chr.1: 171503478 - 171504750 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00727241_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alyref2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019484.4 NP_062357.3 1 1127 133
          GenBank mRNA AK048243.1 - 1 1127 133

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alyref2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          transport
          RNA splicing
          mRNA transport
          nucleotide binding
          nucleic acid binding
          single-stranded DNA binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828236 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337236 AK048243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337236 AK048243.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161139 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337236 AK048243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337236 AK048243.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828212 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161139 - -
          Whole Mouse Genome G4122A A_52_P337236 AK048243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337236 AK048243.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139278 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00749338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00948510 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00629053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01074623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00948515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828242 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20828235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01046181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00486758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01046346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00599460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00534344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01116867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00722436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00629053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00948510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01046345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01060515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351638 - -
          Affymetrix Mouse Gene 1.1 ST Array 10360147 - -
          Affymetrix Mouse Gene 1.1 ST Array 10351636 - -
          Mouse Genome 430 2.0 Array 1422993_PM_s_at - -
          Mouse Genome 430 2.0 Array 1422993_s_at - -
          Affymetrix Mouse Clariom S Array PSR0100013163.mm.1 NM_019484.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013163.mm.1 NM_019484.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013163.mm.1 TC0100001593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013164.mm.1 NM_019484.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013164.mm.1 NM_019484.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013164.mm.1 TC0100001593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013165.mm.1 NM_019484.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013165.mm.1 NM_019484.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013165.mm.1 TC0100001593.mm.1 MTA TC
          Mouse Expression Set 430 1422993_s_at - -
          Mouse Expression Set 430 1422993_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422993_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422993_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 329908 - -
          ABI Mouse Genome Survey Microarray 337648 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000324 - -
          Illumina mouse-6 v1.0 expression beadchip 7000324 - -
          Illumina mouse-6 v1.0 expression beadchip 103520280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697266 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697266 - -
          mouse-6 v1.1 (Illumina) scl17222.1.1_303-S - -
          Illumina Sentrix mouse-6.v1 bead array 103520280 - -
          Illumina Sentrix mouse-6.v1 bead array 7000324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17222.1.1_303-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103520280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2697266 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2697266 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline