Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729099_s1
          See other OLFR738 GE Assays ›
          Gene Symbol:
          Olfr738
          Gene Name:
          olfactory receptor 738
          Gene Aliases:
          MOR106-3
          Chromosome Location:
          Chr.14: 50413546 - 50414481 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729099_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr738
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146420.2 NP_666532.2 1 750 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr738

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146724 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261840 NM_146420.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597370 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146724 - -
          Whole Mouse Genome G4122A A_51_P261840 NM_146420.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261840 NM_146420.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106706 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136648 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11708618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11388353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11831009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11708616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11708605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414476 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414475 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060576 - -
          Illumina mouse-6 v1.0 expression beadchip 3060576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756841 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756841 - -
          mouse-6 v1.1 (Illumina) scl46375.1.130_260-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46375.1.130_260-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2839257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2839257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline