Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731794_g1
          See other POLE3 GE Assays ›
          Gene Symbol:
          Pole3
          Gene Name:
          polymerase (DNA directed), epsilon 3 (p17 subunit)
          Gene Aliases:
          1810034K18Rik, YBL1
          Chromosome Location:
          Chr.4: 62523798 - 62525014 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731794_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pole3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021498.2 NP_067473.2 2 - 3 172 136
          GenBank mRNA AF230806.1 - 2 - 3 172 136
          BC024996.1 - 2 - 3 172 5253060 136
          BC034004.1 - 180 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pole3

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          histone H3 acetylation
          DNA binding
          DNA-directed DNA polymerase activity
          protein binding
          transferase activity
          nucleotidyltransferase activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964956 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 AF230806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 AK007693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 BC024996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 BC034004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 NM_021498.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 AF230806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 AK007693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 BC024996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 BC034004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492087 NM_021498.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80593 AK007693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80593 BC034004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80593 AK007693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80593 BC034004.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168055 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100582 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 AF230806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 AK007693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 BC024996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 BC034004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 NM_021498.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 AF230806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 AK007693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 BC024996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 BC034004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492087 NM_021498.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 AF230806.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 AK007693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 BC024996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 BC034004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 NM_021498.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 AF230806.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 AK007693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 BC024996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 BC034004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492087 NM_021498.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964955 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100582 - -
          Whole Mouse Genome G4122A A_51_P492087 AF230806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 AK007693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 BC024996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 BC034004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 NM_021498.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 AF230806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 AK007693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 BC024996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 BC034004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492087 NM_021498.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80593 AK007693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80593 BC034004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80593 AK007693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80593 BC034004.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122182 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912037 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10513626 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513625 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513628 - -
          Affymetrix HT MG-430 PM Array Plate 1421015_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1452743_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459816_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10513622 - -
          Mouse Genome 430 2.0 Array 1421015_PM_s_at - -
          Mouse Genome 430 2.0 Array 1459816_PM_x_at - -
          Mouse Genome 430 2.0 Array 1421015_s_at - -
          Mouse Genome 430 2.0 Array 1459816_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 59001_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 59001_at - -
          Affymetrix Mouse Clariom S Array PSR0400022934.mm.1 NM_021498.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022934.mm.1 NM_021498.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022934.mm.1 TC0400002792.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022932.mm.1 NM_021498.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022932.mm.1 NM_021498.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022932.mm.1 TC0400002792.mm.1 MTA TC
          Mouse Expression Set 430 1459816_x_at - -
          Mouse Expression Set 430 1421015_s_at - -
          Mouse Expression Set 430 1421015_PM_s_at - -
          Mouse Expression Set 430 1459816_PM_x_at - -
          Murine Genome U74v2 Set 100408_at - -
          Murine Genome U74v2 Set 100408_PM_at - -
          Murine Genome U74v2 Set 162096_at - -
          Murine Genome U74v2 Set 160743_at - -
          Murine Genome U74v2 Set 162096_PM_at - -
          Murine Genome U74v2 Set 160743_PM_at - -
          Mouse Genome 430A 2.0 Array 1459816_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1459816_x_at - -
          Mouse Genome 430A 2.0 Array 1421015_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1421015_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1459816_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1459816_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421015_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452743_at - -
          Affymetrix HT Mouse Genome 430A Array 1421015_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452743_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368438 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270019 - -
          Illumina mouse-6 v1.0 expression beadchip 105290097 - -
          Illumina mouse-6 v1.0 expression beadchip 5270019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247613 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247613 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254553 - -
          mouse-6 v1.1 (Illumina) scl24260.3_379-S - -
          mouse-6 v1.1 (Illumina) scl0059001.2_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 105290097 - -
          Illumina Sentrix mouse-6.v1 bead array 5270019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0059001.2_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270019 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105290097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247613 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2840296 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247613 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2840296 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline