Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731962_s1
          See other OLFR1431 GE Assays ›
          Gene Symbol:
          Olfr1431
          Gene Name:
          olfactory receptor 1431
          Gene Aliases:
          MOR214-5
          Chromosome Location:
          Chr.19: 12209568 - 12210506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731962_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1431
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146414.1 NP_666526.1 1 53 128
          GenBank mRNA BC133710.1 - 1 57 40142241 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1431

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P208416 NM_146414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P208416 NM_146414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151443 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P208416 NM_146414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P208416 BC133710.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P208416 NM_146414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P208416 NM_146414.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P208416 NM_146414.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959892 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151443 - -
          Whole Mouse Genome G4122A A_52_P208416 NM_146414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208416 BC133710.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208416 NM_146414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208416 BC133710.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132466 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101284 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126791 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14511468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14511453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14511461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14511438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14511445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14391613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14142695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461708 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461705 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461707 - -
          Affymetrix Mouse Clariom S Array PSR1900002880.mm.1 NM_146414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002880.mm.1 NM_146414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002880.mm.1 TC1900000239.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652087 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100060500 - -
          Illumina mouse-6 v1.0 expression beadchip 100060500 - -
          Illumina mouse-6 v1.0 expression beadchip 5690368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753911 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753911 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222755 - -
          mouse-6 v1.1 (Illumina) scl0258409.1_48-S - -
          mouse-6 v1.1 (Illumina) GI_33239081-S - -
          Illumina Sentrix mouse-6.v1 bead array 100060500 - -
          Illumina Sentrix mouse-6.v1 bead array 5690368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258409.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690368 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100060500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753911 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2963115 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2963115 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline