Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732037_s1
          See other OLFR919 GE Assays ›
          Gene Symbol:
          Olfr919
          Gene Name:
          olfactory receptor 919
          Gene Aliases:
          MOR171-23
          Chromosome Location:
          Chr.9: 38697429 - 38698376 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732037_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr919
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146440.1 NP_666651.1 1 54 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002064.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr919

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976800 NM_146609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986686 NM_146440.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169109 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986686 NM_146440.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447509 NM_146440.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435687 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169109 - -
          Whole Mouse Genome G4122A A_51_P447509 NM_146440.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07557028 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07557019 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07557015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07557013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07557017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07693369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435697 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584505 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592380 - -
          Affymetrix Mouse Clariom S Array PSR0900017176.mm.1 NM_146440.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017176.mm.1 NM_146440.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017176.mm.1 TC0900002064.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2730704 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2730704 - -
          mouse-6 v1.1 (Illumina) scl36016.1.1_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36016.1.1_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2730704 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2920889 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2920889 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline