Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00772670_g1
          See other APTX GE Assays ›
          Gene Symbol:
          Aptx
          Gene Name:
          aprataxin
          Gene Aliases:
          2410016G21Rik, AA388047, FHA-HIT
          Chromosome Location:
          Chr.4: 40682078 - 40721667 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00772670_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aptx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025444.3 NP_001020615.1 3 - 4 241 88
          NM_025545.4 NP_079821.3 3 - 4 376 88
          GenBank mRNA AK005286.1 - 2 - 3 193 88
          AK077351.1 - 3 - 4 377 88
          AY040780.1 - 3 - 4 387 88
          AY208844.1 - 3 - 4 388 88
          BB860888.1 - 3 - 4 245 88
          BC021872.1 - 3 - 4 234 3994375 88
          BC071213.1 - 3 - 4 228 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002444.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aptx

          Panther Classification:

          Molecular Function -

          damaged DNA-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          DNA ligation
          DNA repair
          double-strand break repair
          cellular response to DNA damage stimulus
          dephosphorylation
          regulation of protein stability
          response to hydrogen peroxide
          nucleic acid phosphodiester bond hydrolysis
          polynucleotide 3' dephosphorylation
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          double-stranded RNA binding
          catalytic activity
          GPI-anchor transamidase activity
          phosphoglycolate phosphatase activity
          hydrolase activity
          DNA 5'-adenosine monophosphate hydrolase activity
          polynucleotide 3'-phosphatase activity
          metal ion binding
          protein N-terminus binding
          phosphoprotein binding
          coenzyme F390-A hydrolase activity
          coenzyme F390-G hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400019684.mm.1 NM_001025444.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019684.mm.1 NM_001025445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019684.mm.1 NM_025545.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019684.mm.1 NM_025545.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019684.mm.1 TC0400002444.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400019683.mm.1 NM_001025444.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019683.mm.1 NM_001025445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019683.mm.1 NM_025545.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019683.mm.1 NM_025545.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400019683.mm.1 TC0400002444.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline