Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00774698_m1
          See other SETMAR GE Assays ›
          Gene Symbol:
          Setmar
          Gene Name:
          SET domain without mariner transposase fusion
          Gene Aliases:
          5830404F24Rik, Etet2
          Chromosome Location:
          Chr.6: 108065045 - 108077127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00774698_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Setmar
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178391.4 NP_848478.2 1 - 2 187 138
          GenBank mRNA AK017895.1 - 1 - 2 138
          BB662956.1 - 1 - 2 188 138
          BC045208.1 - 1 - 2 177 5067523 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001192.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Setmar

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin modification
          methylation
          histone lysine methylation
          methyltransferase activity
          zinc ion binding
          transferase activity
          histone-lysine N-methyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 AK017895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 BC045208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 NM_178391.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 AK017895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 BC045208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339380 NM_178391.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780501 AK036785.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165803 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160778 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 AK017895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 BC045208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 NM_178391.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 AK017895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 BC045208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339380 NM_178391.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165803 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160778 - -
          Whole Mouse Genome G4122A A_51_P339380 AK017895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339380 BC045208.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339380 NM_178391.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339380 AK017895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339380 BC045208.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339380 NM_178391.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122494 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160778 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05408885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05215860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05560758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05156911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05156882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05720332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05874678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05156899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05729294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05156900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05729278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05560755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05225526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05225513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05874674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05676255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05729281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24731368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10540406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540407 - -
          Affymetrix HT MG-430 PM Array Plate 1429389_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10540405 - -
          Mouse Genome 430 2.0 Array 1429389_at - -
          Mouse Genome 430 2.0 Array 1429389_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74729_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74729_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600009188.mm.1 NM_001276356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009188.mm.1 NM_178391.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009188.mm.1 TC0600001192.mm.1 MTA TC
          Mouse Expression Set 430 1429389_PM_at - -
          Mouse Expression Set 430 1429389_at - -
          Affymetrix HT Mouse Genome 430B Array 1429389_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429389_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375317 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237671 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237671 - -
          mouse-6 v1.1 (Illumina) scl29676.3_683-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29676.3_683-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237671 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237671 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline