Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00774982_g1
          See other H2-M10.2 GE Assays ›
          Gene Symbol:
          H2-M10.2
          Gene Name:
          histocompatibility 2, M region locus 10.2
          Gene Aliases:
          4.7H
          Chromosome Location:
          Chr.17: 36284281 - 36286421 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00774982_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-M10.2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177923.1 NP_808591.1 2 - 3 339 100
          GenBank mRNA AF539598.1 - 2 - 3 339 100
          AY211078.1 - 322 100
          BC137985.1 - 2 - 3 408 8861004 100
          BC137987.1 - 2 - 3 408 8861006 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-M10.2

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107947 BC137987.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107947 AY211078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107947 BC137985.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423785 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134923 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107947 BC137987.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107947 AY211078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107947 BC137985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291723 BC137987.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291723 AF539598.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291723 AY211078.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291723 BC137985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291723 NM_177923.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134923 - -
          Whole Mouse Genome G4122A A_52_P291723 BC137987.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291723 AF539598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291723 AY211078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291723 BC137985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291723 NM_177923.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133656 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13551195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13551172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450766 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450762 - -
          Affymetrix Mouse Clariom S Array PSR1700019341.mm.1 NM_177923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019341.mm.1 NM_177923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019341.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019343.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019340.mm.1 NM_177923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019340.mm.1 NM_177923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019340.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019338.mm.1 NM_177923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019338.mm.1 NM_177923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019338.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019342.mm.1 NM_177923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019342.mm.1 NM_177923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019342.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019336.mm.1 TC1700002019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019339.mm.1 NM_177923.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019339.mm.1 NM_177923.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019339.mm.1 TC1700002019.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 433486 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5050148 - -
          Illumina mouse-6 v1.0 expression beadchip 5050148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737429 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737429 - -
          mouse-6 v1.1 (Illumina) scl0333715.4_94-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0333715.4_94-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737429 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737429 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline