Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00775259_g1
          See other CYP2D13 GE Assays ›
          Gene Symbol:
          Cyp2d13
          Gene Name:
          cytochrome P450, family 2, subfamily d, polypeptide 13
          Gene Aliases:
          1300007K12Rik
          Chromosome Location:
          Chr.15: 82636750 - 82642045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00775259_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2d13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003552.1 2 - 3 431 102
          GenBank mRNA BC018344.1 - 2 - 3 411 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001953.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968195 NR_003552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968195 AK004933.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108573 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161934 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968195 NR_003552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968195 AK004933.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161934 - -
          Whole Mouse Genome G4122A A_52_P372062 BC018344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372062 NR_003552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372062 AK004933.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11912810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12207941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11982371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11982366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12435382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11872508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11872509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12435386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11982363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11837169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11968125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11837179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12357426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12207943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30384067 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430891 - -
          Affymetrix HT MG-430 PM Array Plate 1431803_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425365_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430883 - -
          Mouse Genome 430 2.0 Array 1431803_PM_at - -
          Mouse Genome 430 2.0 Array 1431803_at - -
          Mouse Genome 430 2.0 Array 1425365_a_at - -
          Mouse Genome 430 2.0 Array 1425365_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68444_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68444_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500015375.mm.1 NR_003552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015375.mm.1 TC1500001953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015378.mm.1 NR_003552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015378.mm.1 TC1500001953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015374.mm.1 NR_003552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015374.mm.1 TC1500001953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015376.mm.1 NR_003552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015376.mm.1 TC1500001953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015377.mm.1 NR_003552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015377.mm.1 TC1500001953.mm.1 MTA TC
          Mouse Expression Set 430 1425365_a_at - -
          Mouse Expression Set 430 1431803_at - -
          Mouse Expression Set 430 1425365_PM_a_at - -
          Mouse Expression Set 430 1431803_PM_at - -
          Mouse Genome 430A 2.0 Array 1425365_a_at - -
          Mouse Genome 430A 2.0 Array 1425365_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1431803_at - -
          Mouse Genome 430A 2.0 Array 1431803_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1431803_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425365_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425365_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431803_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 772131 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline