Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00775695_mH
          See other CYB5R4 GE Assays ›
          Gene Symbol:
          Cyb5r4
          Gene Name:
          cytochrome b5 reductase 4
          Gene Aliases:
          2810034J18Rik, B5+B5R, C79736, Cb5cb5r, Ncb5or, b5/b5r, b5b5r
          Chromosome Location:
          Chr.9: 87022008 - 87077774 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00775695_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb5r4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024195.2 NP_077157.2 4 - 5 562 116
          GenBank mRNA AF338818.1 - 3 - 4 310 116
          AK012855.1 - 3 - 4 201 116
          AK034878.1 - 4 - 5 506 116
          AK090159.1 - 4 - 5 564 116
          AK135261.1 - 3 - 4 213 116
          AK157312.1 - 4 - 5 542 116
          AK167436.1 - 3 - 4 260 116
          AK169937.1 - 4 - 5 536 116
          AK169995.1 - 4 - 5 533 115
          AK172252.1 - 4 - 5 491 116
          AY321368.1 - 3 - 4 310 116
          AY321369.1 - 3 - 4 310 116
          BC025438.1 - 4 - 5 549 5038563 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001123.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb5r4

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          NADP metabolic process
          superoxide metabolic process
          insulin secretion
          heme metabolic process
          glucose homeostasis
          response to antibiotic
          cell development
          oxidation-reduction process
          NADPH-hemoprotein reductase activity
          cytochrome-b5 reductase activity, acting on NAD(P)H
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
          heme binding
          metal ion binding
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900008819.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008819.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008819.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008809.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008809.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008809.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008811.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008811.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008811.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008818.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008818.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008818.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008812.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008812.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008812.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008813.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008813.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008813.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008814.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008814.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008814.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008816.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008816.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008816.mm.1 TC0900001123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008815.mm.1 NM_024195.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008815.mm.1 NM_024195.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008815.mm.1 TC0900001123.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926362 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline