Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783707_s1
          See other H2-DMB2 GE Assays ›
          Gene Symbol:
          H2-DMb2
          Gene Name:
          histocompatibility 2, class II, locus Mb2
          Gene Aliases:
          H-2Mb2, H2-Mb2
          Chromosome Location:
          Chr.17: 34145416 - 34151095 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783707_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-DMb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010388.4 NP_034518.1 2 150 149
          GenBank mRNA AK154923.1 - 2 254 149
          BC052864.1 - 3 405 149
          BC141105.1 - 2 216 9055717 149
          BC145385.1 - 2 216 9053943 149
          BC145386.1 - 2 216 9053944 149
          BC171960.1 - 2 216 9054246 149
          KJ650224.1 - 2 150 149
          U20314.1 - 1 96 149
          U20315.1 - 1 96 149
          U20316.1 - 1 96 149
          U20317.1 - 1 96 149
          U35334.1 - 2 150 149
          U35335.1 - 2 150 149
          U35336.1 - 2 150 149
          U35338.1 - 2 150 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-DMb2

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          chaperone mediated protein folding requiring cofactor
          protein binding
          MHC class II protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 U35334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 U35335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 NM_010388.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 AK154923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 U35334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 U35335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 NM_010388.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 AK154923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55761 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 U35334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 U35335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 NM_010388.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 AK154923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 BC052864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 U35334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 U35335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 NM_010388.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 AK154923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 U35334.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 U35335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 NM_010388.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 AK154923.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 U35334.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 U35335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 NM_010388.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 AK154923.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152395 - -
          Whole Mouse Genome G4122A A_52_P146877 X90808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146877 X90807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146877 X90800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146877 X90801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146877 X90799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 U35334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 U35335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 NM_010388.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 AK154923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 BC052864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 U35334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 U35335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 NM_010388.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 AK154923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1391 BC052864.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13585530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13477530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13150807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13585535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13336003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13576154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13280020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13493413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13576157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13395716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13176518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13576155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13576567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13576158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13357160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13091613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444240 - -
          Affymetrix HT MG-430 PM Array Plate 1419744_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443687_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1443686_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444236 - -
          Mouse Genome 430 2.0 Array 1443687_x_at - -
          Mouse Genome 430 2.0 Array 1443686_PM_at - -
          Mouse Genome 430 2.0 Array 1419744_PM_at - -
          Mouse Genome 430 2.0 Array 1443687_PM_x_at - -
          Mouse Genome 430 2.0 Array 1443686_at - -
          Mouse Genome 430 2.0 Array 1449580_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449580_s_at - -
          Mouse Genome 430 2.0 Array 1419744_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15000_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15000_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700005846.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005846.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005846.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005847.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005847.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005847.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005842.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005843.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005843.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005843.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005848.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005848.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005848.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005849.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005849.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005849.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005851.mm.1 TC1700002779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005845.mm.1 NM_010388.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005845.mm.1 NM_010388.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005845.mm.1 TC1700002779.mm.1 MTA TC
          Mouse Expression Set 430 1419744_PM_at - -
          Mouse Expression Set 430 1418638_PM_at - -
          Mouse Expression Set 430 1418638_at - -
          Mouse Expression Set 430 1419744_at - -
          Mouse Expression Set 430 1449580_s_at - -
          Mouse Expression Set 430 1449580_PM_s_at - -
          Mouse Expression Set 430 1443687_x_at - -
          Mouse Expression Set 430 1443687_PM_x_at - -
          Mouse Expression Set 430 1443686_PM_at - -
          Mouse Expression Set 430 1443686_at - -
          Murine Genome U74v2 Set 101868_PM_i_at - -
          Murine Genome U74v2 Set 101868_i_at - -
          Mouse Genome 430A 2.0 Array 1418638_PM_at - -
          Mouse Genome 430A 2.0 Array 1418638_at - -
          Mouse Genome 430A 2.0 Array 1419744_PM_at - -
          Mouse Genome 430A 2.0 Array 1419744_at - -
          Mouse Genome 430A 2.0 Array 1449580_s_at - -
          Mouse Genome 430A 2.0 Array 1449580_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419744_at - -
          Affymetrix HT Mouse Genome 430A Array 1419744_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443687_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443687_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443686_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443686_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 344672 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450593 - -
          Illumina mouse-6 v1.0 expression beadchip 6450593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450593 - -
          mouse-6 v1.1 (Illumina) scl015000.6_157-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015000.6_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2979639 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589164 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2979639 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl015000.6_157-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline