Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783915_s1
          See other SERPINB9G GE Assays ›
          Gene Symbol:
          Serpinb9g
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B, member 9g
          Gene Aliases:
          1600002F03Rik, AW545779, NK21B
          Chromosome Location:
          Chr.13: 33484790 - 33496002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783915_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb9g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011455.3 NP_035585.2 7 1260 117
          GenBank mRNA AF425083.1 - 7 1255 117
          AK005394.1 - 7 1260 117
          BC064758.1 - 7 1245 30018322 117
          BC064759.1 - 7 1235 30432705 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb9g

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971419 BC064758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971419 BC064759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971419 NM_011455.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971419 AF425083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971419 AK005394.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971419 BC064758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971419 BC064759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971419 NM_011455.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971419 AF425083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971419 AK005394.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28934115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02660968 - -
          Whole Mouse Genome G4122A A_51_P251687 BC064758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251687 BC064759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251687 NM_011455.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251687 AF425083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251687 AK005394.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P31750 NM_183197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P31750 BC052216.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P31750 U96708.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134510 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119285 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120924 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138638 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404485 - -
          Affymetrix HT MG-430 PM Array Plate 1418422_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404479 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404464 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404472 - -
          Mouse Genome 430 2.0 Array 1418423_PM_s_at - -
          Mouse Genome 430 2.0 Array 1418422_PM_at - -
          Mouse Genome 430 2.0 Array 1418423_s_at - -
          Mouse Genome 430 2.0 Array 1418422_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93806_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93806_at - -
          Mouse Expression Set 430 1418423_s_at - -
          Mouse Expression Set 430 1418423_PM_s_at - -
          Murine Genome U74v2 Set 161115_r_at - -
          Murine Genome U74v2 Set 161115_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1418423_s_at - -
          Mouse Genome 430A 2.0 Array 1418423_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418422_at - -
          Affymetrix HT Mouse Genome 430A Array 1418422_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 923763 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2100398 - -
          Illumina mouse-6 v1.0 expression beadchip 2100398 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002100398 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610470 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610470 - -
          mouse-6 v1.1 (Illumina) scl0020709.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 2100398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0020709.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2100398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2610470 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2610470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline