Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00787910_sH
          See other ORMDL3 GE Assays ›
          Gene Symbol:
          Ormdl3
          Gene Name:
          ORM1-like 3 (S. cerevisiae)
          Gene Aliases:
          2810011N17Rik, AI315223
          Chromosome Location:
          Chr.11: 98581294 - 98587245 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00787910_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ormdl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025661.4 NP_079937.1 4 848 118
          GenBank mRNA AK009621.1 - 4 849 118
          AK012718.1 - 4 818 118
          AK087497.1 - 4 817 118
          AK154085.1 - 4 840 118
          BC026412.1 - 2 523 118
          BC046594.1 - 4 786 6308578 118

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ormdl3

          Gene Ontology Categories:

          Function(s) Process(es)

          ceramide metabolic process
          cellular sphingolipid homeostasis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057359 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059347 BC046594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059347 AK087497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059347 AK154085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059347 BC026412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059347 NM_025661.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502530 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100233 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059347 BC046594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059347 AK087497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059347 AK154085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059347 BC026412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059347 NM_025661.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28057348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07458800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100233 - -
          Whole Mouse Genome G4122A A_51_P221460 BC046594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221460 AK087497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221460 AK154085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221460 BC026412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221460 NM_025661.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138273 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685130 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09568008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09685144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057359 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28057354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28057337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09568008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09028307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09731887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28057355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390687 - -
          Affymetrix HT MG-430 PM Array Plate 1419450_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10390685 - -
          Mouse Genome 430 2.0 Array 1419450_at - -
          Mouse Genome 430 2.0 Array 1419450_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66612_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66612_at - -
          Affymetrix Mouse Clariom S Array PSR1100033908.mm.1 TC1100003683.mm.1 MTA TC
          Mouse Expression Set 430 1419450_at - -
          Mouse Expression Set 430 1419450_PM_at - -
          Murine Genome U74v2 Set 98065_at - -
          Murine Genome U74v2 Set 98065_PM_at - -
          Mouse Genome 430A 2.0 Array 1419450_PM_at - -
          Mouse Genome 430A 2.0 Array 1419450_at - -
          Affymetrix HT Mouse Genome 430A Array 1419450_at - -
          Affymetrix HT Mouse Genome 430A Array 1419450_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894065 - -
          ABI Mouse Genome Survey Microarray 903558 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340711 - -
          Illumina mouse-6 v1.0 expression beadchip 1340711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712280 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712280 - -
          mouse-6 v1.1 (Illumina) scl066612.1_144-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066612.1_144-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066612.1_144-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline