Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00802114_m1
          See other APBB2 GE Assays ›
          Gene Symbol:
          Apbb2
          Gene Name:
          amyloid beta (A4) precursor protein-binding, family B, member 2
          Gene Aliases:
          2310007D03Rik, FE65L1, Rirl1, TR2L, Zfra
          Chromosome Location:
          Chr.5: 66298725 - 66618817 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00802114_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apbb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001201413.1 NP_001188342.1 10 - 11 1970 59
          NM_001201414.1 NP_001188343.1 9 - 10 1973 59
          NM_001201415.1 NP_001188344.1 9 - 10 1907 59
          NM_009686.2 NP_033816.1 10 - 11 1970 59
          GenBank mRNA AK004792.1 - 10 - 11 1824 59
          AK153260.1 - 9 - 10 1939 59
          AK167649.1 - 10 - 11 1835 59
          AK169959.1 - 9 - 10 1848 59
          BC042797.1 - 1 - 2 115 59
          BC076587.1 - 9 - 10 1753 30550087 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002508.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apbb2

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of transcription, DNA-templated
          cell cycle arrest
          axon guidance
          actin filament-based movement
          extracellular matrix organization
          negative regulation of cell growth
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          protein stabilization
          beta-amyloid binding
          protein binding
          transcription factor binding
          histone acetyltransferase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_001201415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 NM_009686.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022400.mm.1 TC0500002508.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline