Search
Search
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301737.1 | NP_001288666.1 | 11 - 12 | 1543 | 71 | |
| NM_010065.3 | NP_034195.2 | 11 - 12 | 1543 | 71 | ||
| NR_125959.1 | 11 - 12 | 1543 | 71 | |||
| GenBank mRNA | AK011651.1 | - | 11 - 12 | 1510 | 71 | |
| AK144142.1 | - | 11 - 12 | 1494 | 71 | ||
| AK220483.1 | - | 11 - 12 | 1485 | 71 | ||
| BC034679.1 | - | 11 - 12 | 1497 | 4506865 | 71 | |
| BC058623.1 | - | 11 - 12 | 1477 | 5706803 | 71 | |
| BQ179370.1 | - | 5 - 6 | 468 | 71 | ||
| L31395.1 | - | 11 - 12 | 1440 | 71 | ||
| L31396.1 | - | 6 - 7 | 712 | 71 | ||
| L31397.1 | - | 11 - 12 | 1496 | 71 | ||
Set Membership: |
|