Search
Search

Adgre1| Gene Symbol: Adgre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010130.4 | NP_034260.1 | 14 - 15 | 1794 | 111 | |
| GenBank mRNA | AK133202.1 | - | 14 - 15 | 1770 | 111 | |
| AK149898.1 | - | 14 - 15 | 1779 | 111 | ||
| AK151303.1 | - | 14 - 15 | 1766 | 111 | ||
| AK151682.1 | - | 14 - 15 | 1786 | 111 | ||
| AK155862.1 | - | 14 - 15 | 1745 | 111 | ||
| BC075688.1 | - | 14 - 15 | 1766 | 30609706 | 111 | |
| U66888.1 | - | 14 - 15 | 1781 | 111 | ||
| X93328.1 | - | 14 - 15 | 1790 | 111 | ||
Set Membership: |
|