Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00804095_m1
          See other RAP1B GE Assays ›
          Gene Symbol:
          Rap1b
          Gene Name:
          RAS related protein 1b
          Gene Aliases:
          2810443E11Rik
          Chromosome Location:
          Chr.10: 117814597 - 117845974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00804095_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rap1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024457.2 NP_077777.1 2 - 3 227 114
          GenBank mRNA AK143910.1 - 2 - 3 210 114
          AK153870.1 - 2 - 3 184 114
          AK163242.1 - 2 - 3 291 114
          AK167548.1 - 2 - 3 183 114
          AK178034.1 - 2 - 3 100 114
          BC033382.1 - 2 - 3 167 114
          BC052480.1 - 2 - 3 227 114
          BC094066.1 - 2 - 3 167 114
          BC147390.1 - 2 - 3 99 9056464 114
          BC147391.1 - 2 - 3 99 9056441 114
          BC147700.1 - 1 - 2 85 9087459 114
          BC147713.1 - 1 - 2 85 9087472 114
          BU848028.1 - 232 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002945.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rap1b

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          metabolic process
          cell proliferation
          Rap protein signal transduction
          negative regulation of calcium ion-dependent exocytosis
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          regulation of cell junction assembly
          regulation of establishment of cell polarity
          negative regulation of synaptic vesicle exocytosis
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          protein complex binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000022332.mm.1 NM_024457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022332.mm.1 NM_024457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022332.mm.1 TC1000002945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022329.mm.1 NM_024457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022329.mm.1 NM_024457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022329.mm.1 TC1000002945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022328.mm.1 NM_024457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022328.mm.1 NM_024457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022328.mm.1 TC1000002945.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022333.mm.1 NM_024457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022333.mm.1 NM_024457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022333.mm.1 TC1000002945.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 705625 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline