Search
Search

Fbxw9| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026791.2 | NP_081067.2 | 3 - 4 | 687 | 64 | |
| GenBank mRNA | AK082085.1 | - | 3 - 4 | 1091 | 64 | |
| AK156709.1 | - | 3 - 4 | 753 | 64 | ||
| BC043658.1 | - | 3 - 4 | 648 | 64 | ||
| BC053694.1 | - | 1 - 2 | 103 | 64 | ||
| BC144773.1 | - | 3 - 4 | 1053 | 64 | ||
| BC145661.1 | - | 3 - 4 | 1053 | 64 | ||
| CF197934.1 | - | 1 - 2 | 91 | 64 | ||
Set Membership: |
|