Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805189_g1
          See other FBXO2 GE Assays ›
          Gene Symbol:
          Fbxo2
          Gene Name:
          F-box protein 2
          Gene Aliases:
          FBG1, FBX2, Fbs1, Fbs2, NFB42, Prpl4
          Chromosome Location:
          Chr.4: 148160226 - 148166417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805189_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176848.1 NP_789818.1 5 - 6 778 84
          GenBank mRNA AK140175.1 - 5 - 6 822 84
          BC027053.1 - 4 - 5 731 84
          BC046586.1 - 5 - 6 778 6487759 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo2

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          glycoprotein catabolic process
          negative regulation of cell proliferation
          protein ubiquitination
          ER-associated ubiquitin-dependent protein catabolic process
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          regulation of protein ubiquitination
          beta-amyloid binding
          glycoprotein binding
          protein binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392783 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392757 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392784 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392758 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092296 NM_176848.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092296 AK140175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092296 BC027053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092296 BC046586.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177724 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137239 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092296 NM_176848.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092296 AK140175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092296 BC027053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092296 BC046586.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114472 NM_176848.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114472 AK140175.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114472 BC027053.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114472 BC046586.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392771 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01013359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137239 - -
          Whole Mouse Genome G4122A A_52_P36190 NM_176848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P36190 AK140175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P36190 BC027053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P36190 BC046586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580912 NM_176848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580912 AK140175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580912 BC027053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580912 BC046586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114472 NM_176848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114472 AK140175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114472 BC027053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114472 BC046586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113294 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03904979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03904977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03904976 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392758 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392784 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392783 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392757 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04085862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03506738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03904980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04085850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03904983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03904977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03249569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03249570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510302 - -
          Affymetrix HT MG-430 PM Array Plate 1427004_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510299 - -
          Mouse Genome 430 2.0 Array 1427004_at - -
          Mouse Genome 430 2.0 Array 1427004_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230904_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230904_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400015723.mm.1 NM_176848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015723.mm.1 NM_176848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015723.mm.1 TC0400001926.mm.1 MTA TC
          Mouse Expression Set 430 1427004_at - -
          Mouse Expression Set 430 1427004_PM_at - -
          Mouse Genome 430A 2.0 Array 1427004_at - -
          Mouse Genome 430A 2.0 Array 1427004_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427004_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427004_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 655571 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6510091 - -
          Illumina mouse-6 v1.0 expression beadchip 6510091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627556 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627556 - -
          mouse-6 v1.1 (Illumina) scl24708.6.1_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24708.6.1_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2822825 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2822825 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24708.6.1_98-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline