Search
Search
| Gene Symbol: Mypn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182992.2 | NP_892037.2 | 15 - 16 | 3382 | 60 | |
| GenBank mRNA | AK220521.1 | - | 15 - 16 | 3382 | 60 | |
| BC052872.1 | - | 1 - 2 | 50 | 60 | ||
Set Membership: |
|