Search
Search

Btbd16| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081038.2 | NP_001074507.2 | 4 - 5 | 634 | 54 | |
| GenBank mRNA | AK087912.1 | - | 4 - 5 | 636 | 54 | |
| BB559682.2 | - | 2 - 3 | 193 | 54 | ||
Set Membership: |
|