Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00812390_m1
          See other NF1 GE Assays ›
          Gene Symbol:
          Nf1
          Gene Name:
          neurofibromatosis 1
          Gene Aliases:
          AW494271, E030030H24Rik, Nf-1
          Chromosome Location:
          Chr.11: 79335988 - 79581609 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00812390_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010897.2 NP_035027.1 12 - 13 1497 93
          GenBank mRNA AK085050.1 - 13 - 14 1503 93
          L10370.1 - 11 - 12 1342 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001176.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nf1

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          osteoblast differentiation
          metanephros development
          response to hypoxia
          liver development
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          regulation of cell-matrix adhesion
          negative regulation of cell-matrix adhesion
          negative regulation of protein kinase activity
          cell communication
          signal transduction
          Ras protein signal transduction
          negative regulation of neuroblast proliferation
          brain development
          peripheral nervous system development
          heart development
          skeletal muscle tissue development
          negative regulation of cell proliferation
          visual learning
          extrinsic apoptotic signaling pathway via death domain receptors
          regulation of gene expression
          Schwann cell development
          phosphatidylinositol 3-kinase signaling
          negative regulation of angiogenesis
          spinal cord development
          amygdala development
          forebrain astrocyte development
          neural tube development
          cerebral cortex development
          myelination in peripheral nervous system
          actin cytoskeleton organization
          extracellular matrix organization
          collagen fibril organization
          adrenal gland development
          negative regulation of cell migration
          regulation of synaptic transmission, GABAergic
          cellular response to heat
          negative regulation of Rac protein signal transduction
          wound healing
          regulation of cell proliferation
          negative regulation of transcription factor import into nucleus
          positive regulation of apoptotic process
          regulation of GTPase activity
          negative regulation of MAP kinase activity
          regulation of MAPK cascade
          negative regulation of MAPK cascade
          pigmentation
          positive regulation of neuron apoptotic process
          regulation of blood vessel endothelial cell migration
          positive regulation of GTPase activity
          regulation of bone resorption
          regulation of neuron differentiation
          negative regulation of osteoclast differentiation
          regulation of glial cell differentiation
          positive regulation of adenylate cyclase activity
          regulation of angiogenesis
          negative regulation of Ras protein signal transduction
          negative regulation of neurotransmitter secretion
          negative regulation of fibroblast proliferation
          regulation of long-term neuronal synaptic plasticity
          sympathetic nervous system development
          camera-type eye morphogenesis
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          smooth muscle tissue development
          artery morphogenesis
          forebrain morphogenesis
          cognition
          gamma-aminobutyric acid secretion, neurotransmission
          glutamate secretion, neurotransmission
          observational learning
          regulation of long-term synaptic potentiation
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          GTPase activator activity
          protein binding
          microtubule binding
          lipid binding
          phosphatidylethanolamine binding
          phosphatidylcholine binding
          syndecan binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100010973.mm.1 NM_010897.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010973.mm.1 NM_010897.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010973.mm.1 TC1100001176.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline