| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800001066.mm.1 |
NM_181564.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001066.mm.1 |
NM_181564.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001066.mm.1 |
TC1800000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001065.mm.1 |
NM_181564.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001065.mm.1 |
NM_181564.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001065.mm.1 |
TC1800000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001064.mm.1 |
NM_181564.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001064.mm.1 |
NM_181564.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001064.mm.1 |
TC1800000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001067.mm.1 |
NM_181564.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001067.mm.1 |
NM_181564.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001067.mm.1 |
TC1800000155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001063.mm.1 |
NM_181564.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001063.mm.1 |
NM_181564.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001063.mm.1 |
TC1800000155.mm.1 |
MTA TC |