Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00812905_m1
          See other RNF20 GE Assays ›
          Gene Symbol:
          Rnf20
          Gene Name:
          ring finger protein 20
          Gene Aliases:
          4833430L21Rik, AW540162, C79397, mKIAA4116
          Chromosome Location:
          Chr.4: 49632011 - 49656886 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00812905_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163263.1 NP_001156735.1 5 - 6 1103 103
          NM_182999.2 NP_892044.1 6 - 7 950 103
          GenBank mRNA AK048862.1 - 5 - 6 1062 103
          AK051278.1 - 5 - 6 1062 103
          AK053783.1 - 6 - 7 938 103
          AK082529.1 - 5 - 6 1007 103
          AK163905.1 - 2 - 3 316 103
          AK220492.1 - 5 - 6 1019 103
          BC004593.1 - 1 - 2 177 103
          BC052482.1 - 6 - 7 889 6308280 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000555.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf20

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          histone monoubiquitination
          chromatin modification
          histone ubiquitination
          negative regulation of cell migration
          positive regulation of histone methylation
          histone H2B ubiquitination
          positive regulation of transcription, DNA-templated
          negative regulation of mRNA polyadenylation
          positive regulation of histone H2B ubiquitination
          p53 binding
          chromatin binding
          transcription coactivator activity
          mRNA 3'-UTR binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          ubiquitin protein ligase binding
          histone binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400004072.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004072.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004072.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004072.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004072.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004066.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004066.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004066.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004066.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004066.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004070.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004070.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004070.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004070.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004070.mm.1 TC0400000555.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline