| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_001164559.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_001164559.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_001164560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_001164560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_198020.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
NM_198020.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007245.mm.1 |
TC0800003233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_001164559.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_001164559.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_001164560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_001164560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_198020.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
NM_198020.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007248.mm.1 |
TC0800003233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_001164559.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_001164559.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_001164560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_001164560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_198020.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
NM_198020.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800007246.mm.1 |
TC0800003233.mm.1 |
MTA TC |