Search
Search

Luzp2| Gene Symbol: Luzp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178705.5 | NP_848820.4 | 7 - 8 | 1235 | 69 | |
| GenBank mRNA | AK032204.1 | - | 7 - 8 | 892 | 69 | |
| AK034077.1 | - | 7 - 8 | 853 | 69 | ||
| AK034240.1 | - | 7 - 8 | 887 | 69 | ||
| AK082126.1 | - | 7 - 8 | 983 | 69 | ||
| AY164459.1 | - | 7 - 8 | 1232 | 69 | ||
| BC075646.1 | - | 7 - 8 | 865 | 30620475 | 69 | |
Set Membership: |
|