Search
Search
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008568.2 | NP_032594.1 | 7 - 8 | 973 | 215 | |
| GenBank mRNA | AK076059.1 | - | 3 - 4 | 423 | 215 | |
| AK077766.1 | - | 4 - 5 | 536 | 214 | ||
| AK132748.1 | - | 1534 | 215 | |||
| AK133091.1 | - | 7 - 8 | 969 | 215 | ||
| AK144980.1 | - | 7 - 8 | 985 | 215 | ||
| AK149913.1 | - | 7 - 8 | 967 | 213 | ||
| AK150059.1 | - | 7 - 8 | 970 | 215 | ||
| AK150066.1 | - | 7 - 8 | 967 | 213 | ||
| AK154052.1 | - | 7 - 8 | 970 | 215 | ||
| BC023232.1 | - | 7 - 8 | 949 | 215 | ||
| BC065164.1 | - | 7 - 8 | 957 | 6829737 | 215 | |
| BC066024.1 | - | 7 - 8 | 912 | 6829337 | 215 | |
| D26091.1 | - | 7 - 8 | 973 | 215 | ||
Set Membership: |
|