Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833969_s1
          See other IFNA4 GE Assays ›
          Gene Symbol:
          Ifna4
          Gene Name:
          interferon alpha 4
          Gene Aliases:
          Ifa4
          Chromosome Location:
          Chr.4: 88841816 - 88842459 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833969_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010504.2 NP_034634.1 1 304 202
          GenBank mRNA BC116217.2 - 1 259 40091191 202
          BC119349.1 - 1 304 8734098 202
          BC119351.1 - 1 304 8734100 202

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna4

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          defense response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          defense response to virus
          cytokine activity
          cytokine receptor binding
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC116217.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC119349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 NM_010504.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC119351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC116217.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC119349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 NM_010504.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355829 BC119351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC116217.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC119349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 NM_010504.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC119351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC116217.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC119349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 NM_010504.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355829 BC119351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC116217.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC119349.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 NM_010504.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC119351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC116217.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC119349.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 NM_010504.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355829 BC119351.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092492 - -
          Whole Mouse Genome G4122A A_51_P355829 BC116217.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 BC119349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 NM_010504.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 BC119351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 BC116217.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 BC119349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 NM_010504.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355829 BC119351.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108977 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03707506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505891 - -
          Affymetrix HT MG-430 PM Array Plate 1422408_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505890 - -
          Mouse Genome 430 2.0 Array 1422408_at - -
          Mouse Genome 430 2.0 Array 1422408_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15967_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15967_at - -
          Affymetrix Mouse Clariom S Array PSR0400006108.mm.1 NM_010504.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006108.mm.1 NM_010504.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006108.mm.1 TC0400000908.mm.1 MTA TC
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422408_PM_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1422408_at - -
          Murine Genome U74v2 Set 101789_i_at - -
          Murine Genome U74v2 Set 101790_f_at - -
          Murine Genome U74v2 Set 101789_PM_i_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 101790_PM_f_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1422408_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422408_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422408_at - -
          Affymetrix HT Mouse Genome 430A Array 1422408_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375730 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1940025 - -
          Illumina mouse-6 v1.0 expression beadchip 103130064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216457 - -
          mouse-6 v1.1 (Illumina) scl015967.1_72-S - -
          mouse-6 v1.1 (Illumina) GI_6754293-S - -
          Illumina Sentrix mouse-6.v1 bead array 103130064 - -
          Illumina Sentrix mouse-6.v1 bead array 1940025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015967.1_72-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103130064 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline