| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086236 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086234 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086235 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081925 |
AK030965.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2081925 |
NM_013843.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402110 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360248 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186637 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642197 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
108832 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520612 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114557 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
258478 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341693 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
495807 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
60903 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
815242 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
425594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
169848 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
AF159549.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
AK009260.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
AK032324.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
AK156205.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
BC020364.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
BC056192.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479335 |
NM_013745.5 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
950569 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
953741 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242364 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
317394 |
X69859.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
771490 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
320209 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
125491 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809785 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746346 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861850 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521808 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551922 |
AK009203.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551922 |
AK017714.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551922 |
AK159897.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551922 |
BC027311.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551922 |
NM_025857.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
781298 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
110364 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
92044 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151077 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153059 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
169396 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
138873 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16098 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P152255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P150550 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081925 |
AK030965.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2081925 |
NM_013843.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2126585 |
BC023798.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2126585 |
AK090292.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086230 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086241 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086208 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086236 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086237 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086233 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086232 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086229 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086226 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086228 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086217 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086219 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086235 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086240 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086248 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086225 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086247 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086249 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086214 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086220 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086231 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086222 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086238 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086212 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086210 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086234 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31086223 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08734160 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P152255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P150550 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03290789 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08734172 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03290802 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08734094 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P314077 |
BC046328.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P314077 |
AK148857.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P314077 |
BC020362.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P314077 |
AK030965.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P314077 |
NM_013843.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P415735 |
BC046328.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P415735 |
AK148857.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P415735 |
BC020362.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P415735 |
AK030965.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P415735 |
NM_013843.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110921 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134970 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08734094 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P152255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P150550 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08734172 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13433898 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13433893 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13264565 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13264563 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13264564 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13433896 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13433892 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086236 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086235 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31086234 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086359 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086349 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086313 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086209 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086258 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086277 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086337 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086273 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086331 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086357 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086260 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086296 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086304 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086314 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086319 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086297 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086275 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086355 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086353 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086343 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086255 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086249 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086228 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086292 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086253 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086251 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086339 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086291 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086234 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31086271 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086230 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086222 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086237 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086252 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086258 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086208 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13490837 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086207 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086249 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086199 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086260 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086228 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086248 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086240 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086203 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086218 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086261 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086238 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086259 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086212 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13433896 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086229 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086214 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086202 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086219 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086210 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13490832 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086242 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086201 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086231 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13490833 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086232 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13280679 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13294453 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13274190 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086209 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086245 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086241 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086225 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13469265 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086236 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086255 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31086226 |
- |
- |