Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837090_s1
          See other OLFR99 GE Assays ›
          Gene Symbol:
          Olfr99
          Gene Name:
          olfactory receptor 99
          Gene Aliases:
          MOR156-1
          Chromosome Location:
          Chr.17: 37279501 - 37280418 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837090_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146515.2 NP_666726.1 1 845 109
          GenBank mRNA BC106777.1 - 1 845 40040271 109
          BC106778.1 - 1 845 40040272 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr99

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103170 BC106777.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159178 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103170 BC106777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288951 NM_146515.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288951 BC106777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P288951 BC106778.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168178 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159178 - -
          Whole Mouse Genome G4122A A_52_P288951 NM_146515.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288951 BC106777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288951 BC106778.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13395342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13146604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13146597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13361767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13395327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13395334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13497092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450873 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450872 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 899782 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230728 - -
          mouse-6 v1.1 (Illumina) scl49931.1.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49931.1.1_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2811562 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2811562 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230728 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline