Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838045_s1
          See other OLFR799 GE Assays ›
          Gene Symbol:
          Olfr799
          Gene Name:
          olfactory receptor 799
          Gene Aliases:
          MOR114-2
          Chromosome Location:
          Chr.10: 129647130 - 129648065 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838045_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr799
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146927.1 NP_667138.1 1 141 237

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr799

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325664 NM_146927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325664 NM_146927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325664 NM_146927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325664 NM_146927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325664 NM_146927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325664 NM_146927.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546642 - -
          Whole Mouse Genome G4122A A_51_P325664 NM_146927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325664 NM_146927.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123174 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08931060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08798170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08798162 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08798167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08798172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08931062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08798164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367512 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367511 - -
          Affymetrix Mouse Clariom S Array PSR1000012045.mm.1 NM_146927.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012045.mm.1 NM_146927.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012045.mm.1 TC1000001648.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903071 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150309 - -
          Illumina mouse-6 v1.0 expression beadchip 4150309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257470 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257470 - -
          mouse-6 v1.1 (Illumina) scl0258929.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258929.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257470 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3001636 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257470 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3001636 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline