Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840109_m1
          See other HSD17B10 GE Assays ›
          Gene Symbol:
          Hsd17b10
          Gene Name:
          hydroxysteroid (17-beta) dehydrogenase 10
          Gene Aliases:
          17bHSD10, Ads9, ERAB, Hadh2
          Chromosome Location:
          Chr.X: 152001896 - 152004442 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840109_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsd17b10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016763.2 NP_058043.3 2 - 3 210 78
          GenBank mRNA AF233685.1 - 2 - 3 210 78
          AK002368.1 - 2 - 3 205 78
          AK013340.1 - 2 - 3 206 78
          AK077735.1 - 2 - 3 204 78
          AK088685.1 - 2 - 3 226 78
          BC027517.1 - 2 - 3 210 3376098 78
          U96116.1 - 2 - 3 311 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001533.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsd17b10

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          metabolic process
          protein homotetramerization
          oxidation-reduction process
          beta-amyloid binding
          3-hydroxyacyl-CoA dehydrogenase activity
          estradiol 17-beta-dehydrogenase activity
          steroid binding
          oxidoreductase activity
          acetoacetyl-CoA reductase activity
          testosterone dehydrogenase [NAD(P)] activity
          estrogen receptor binding
          identical protein binding
          poly(A) RNA binding
          3-hydroxy-2-methylbutyryl-CoA dehydrogenase activity
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042698 AK088685.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042698 BC027517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042698 AK013340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042698 AK077735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042698 U96116.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486064 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127647 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042698 AK088685.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042698 BC027517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042698 AK013340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042698 AK077735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042698 U96116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366960 AK088685.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366960 BC027517.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366960 AK013340.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366960 AK077735.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366960 U96116.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32747510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127647 - -
          Whole Mouse Genome G4122A A_51_P366960 AK088685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366960 BC027517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366960 AK013340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366960 AK077735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366960 U96116.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132534 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127647 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15058182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14886957 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15058181 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14886958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32747501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14774131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14886955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15058187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15039214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15058186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15081321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32747485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10602596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602598 - -
          Affymetrix HT MG-430 PM Array Plate 1438391_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448286_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10602592 - -
          Mouse Genome 430 2.0 Array 1438391_x_at - -
          Mouse Genome 430 2.0 Array 1438391_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448286_at - -
          Mouse Genome 430 2.0 Array 1448286_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15108_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15108_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00009706.mm.1 NM_016763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009706.mm.1 NM_016763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009706.mm.1 TC0X00001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009703.mm.1 NM_016763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009703.mm.1 NM_016763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009703.mm.1 TC0X00001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009702.mm.1 NM_016763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009702.mm.1 NM_016763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009702.mm.1 TC0X00001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009708.mm.1 NM_016763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009708.mm.1 NM_016763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009708.mm.1 TC0X00001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009709.mm.1 NM_016763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009709.mm.1 NM_016763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009709.mm.1 TC0X00001533.mm.1 MTA TC
          Mouse Expression Set 430 1438391_x_at - -
          Mouse Expression Set 430 1448286_PM_at - -
          Mouse Expression Set 430 1438391_PM_x_at - -
          Mouse Expression Set 430 1448286_at - -
          Murine Genome U74v2 Set 101045_PM_at - -
          Murine Genome U74v2 Set 101045_at - -
          Murine Genome U74v2 Set 169231_PM_at - -
          Murine Genome U74v2 Set 169231_at - -
          Mouse Genome 430A 2.0 Array 1438391_x_at - -
          Mouse Genome 430A 2.0 Array 1438391_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448286_at - -
          Mouse Genome 430A 2.0 Array 1448286_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448286_at - -
          Affymetrix HT Mouse Genome 430A Array 1438391_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438391_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448286_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 426545 - -
          ABI Mouse Genome Survey Microarray 876577 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238614 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238614 - -
          mouse-6 v1.1 (Illumina) scl54548.7.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54548.7.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238614 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238614 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54548.7.1_64-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline