Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840143_g1
          See other SLC35G3 GE Assays ›
          Gene Symbol:
          Slc35g3
          Gene Name:
          solute carrier family 35, member G3
          Gene Aliases:
          Amac1
          Chromosome Location:
          Chr.11: 69759884 - 69761844 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840143_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc35g3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019871.2 NP_063924.2 1 - 2 233 69
          GenBank mRNA AF016712.1 - 1 - 2 82 69
          AK077181.1 - 1 - 2 358 69
          BC137893.1 - 1 - 2 113 8860912 69
          BC137894.1 - 1 - 2 113 8860913 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003062.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc35g3

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896953 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896952 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896954 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127313 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119926 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365704 AF016712.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896947 - -
          Whole Mouse Genome G4122A A_51_P365704 AF016712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365704 AF016712.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126739 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09376142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09376144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09500610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896952 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896953 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27896954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10387623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387622 - -
          Affymetrix HT MG-430 PM Array Plate 1418958_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10387620 - -
          Mouse Genome 430 2.0 Array 1418958_at - -
          Mouse Genome 430 2.0 Array 1418958_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56293_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56293_at - -
          Affymetrix Mouse Clariom S Array PSR1100028091.mm.1 NM_019871.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028091.mm.1 NM_019871.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028091.mm.1 TC1100003062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028092.mm.1 NM_019871.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028092.mm.1 NM_019871.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028092.mm.1 TC1100003062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028093.mm.1 NM_019871.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028093.mm.1 NM_019871.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028093.mm.1 TC1100003062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028094.mm.1 NM_019871.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028094.mm.1 NM_019871.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028094.mm.1 TC1100003062.mm.1 MTA TC
          Mouse Expression Set 430 1418958_PM_at - -
          Mouse Expression Set 430 1418958_at - -
          Murine Genome U74v2 Set 93386_PM_at - -
          Murine Genome U74v2 Set 93386_at - -
          Mouse Genome 430A 2.0 Array 1418958_at - -
          Mouse Genome 430A 2.0 Array 1418958_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418958_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418958_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 847385 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 940270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225521 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225521 - -
          mouse-6 v1.1 (Illumina) scl40018.2_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 940270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40018.2_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225521 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225521 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline