Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840165_g1
          See other FGF21 GE Assays ›
          Gene Symbol:
          Fgf21
          Gene Name:
          fibroblast growth factor 21
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 45613890 - 45619350 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840165_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020013.4 NP_064397.1 1 - 2 425 78
          GenBank mRNA AB025718.1 - 1 - 2 241 78
          AK007574.1 - 1 - 2 425 78
          BC049592.1 - 1 - 2 424 6774352 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003013.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf21

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of glucose import
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          receptor binding
          fibroblast growth factor receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 NM_020013.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 AK007574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 NM_020013.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 AK007574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385232 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128285 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 NM_020013.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 AK007574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 BC049592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 NM_020013.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 AK007574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315925 NM_020013.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315925 AB025718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315925 AK007574.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315925 BC049592.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 NM_020013.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 AK007574.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 NM_020013.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 AK007574.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128285 - -
          Whole Mouse Genome G4122A A_51_P315925 NM_020013.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315925 AB025718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315925 AK007574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315925 BC049592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 NM_020013.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 AK007574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 BC049592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 NM_020013.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 AK007574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P235347 BC049592.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100395 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138111 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06388543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05904172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06388547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06388543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06323714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06323696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06323697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05904178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563347 - -
          Affymetrix HT MG-430 PM Array Plate 1422916_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563344 - -
          Mouse Genome 430 2.0 Array 1422916_at - -
          Mouse Genome 430 2.0 Array 1422916_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56636_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56636_at - -
          Affymetrix Mouse Clariom S Array PSR0700027348.mm.1 NM_020013.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027348.mm.1 NM_020013.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027348.mm.1 TC0700003013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027346.mm.1 NM_020013.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027346.mm.1 NM_020013.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027346.mm.1 TC0700003013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027347.mm.1 NM_020013.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027347.mm.1 NM_020013.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027347.mm.1 TC0700003013.mm.1 MTA TC
          Mouse Expression Set 430 1422916_PM_at - -
          Mouse Expression Set 430 1422916_at - -
          Murine Genome U74v2 Set 165847_PM_i_at - -
          Murine Genome U74v2 Set 165847_i_at - -
          Mouse Genome 430A 2.0 Array 1422916_PM_at - -
          Mouse Genome 430A 2.0 Array 1422916_at - -
          Affymetrix HT Mouse Genome 430A Array 1422916_at - -
          Affymetrix HT Mouse Genome 430A Array 1422916_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 860625 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3780377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710698 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710698 - -
          mouse-6 v1.1 (Illumina) scl31369.3.1_132-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31369.3.1_132-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710698 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710698 - -

          Back To Top
           

          Citations (1)

          Fibroblast growth factor 21 corrects obesity in mice.
          Authors
          Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
          Journal
          Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 UTC 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline