Search
Search

Skint3| Gene Symbol: Skint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177578.4 | NP_808246.1 | 4 - 5 | 916 | 146 | |
| GenBank mRNA | AK040383.1 | - | 4 - 5 | 917 | 146 | |
| EF494895.1 | - | 4 - 5 | 740 | 146 | ||
Set Membership: |
|