Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844064_s1
          See other ATOH7 GE Assays ›
          Gene Symbol:
          Atoh7
          Gene Name:
          atonal homolog 7 (Drosophila)
          Gene Aliases:
          Math5, bHLHa13
          Chromosome Location:
          Chr.10: 63098020 - 63101284 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844064_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atoh7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016864.1 NP_058560.1 1 346 168
          GenBank mRNA AF071223.1 - 1 346 168
          BC092234.1 - 1 699 30940279 168

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atoh7

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          neural retina development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          nervous system development
          circadian rhythm
          entrainment of circadian clock
          optic nerve development
          cell differentiation
          DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189395 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189394 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020760 NM_016864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020760 AF071223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020760 BC092234.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020760 NM_016864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020760 AF071223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020760 BC092234.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27189384 - -
          Whole Mouse Genome G4122A A_52_P684087 BC092234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P684087 BC092234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152102 NM_016864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152102 AF071223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152102 BC092234.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08580962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555458 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555474 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189394 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189395 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08339263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08277579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08300452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08580962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08812719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08222984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08918429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08555468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08555464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08555451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08918423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189353 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10363640 - -
          Affymetrix HT MG-430 PM Array Plate 1450796_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422929_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10363639 - -
          Mouse Genome 430 2.0 Array 1422929_s_at - -
          Mouse Genome 430 2.0 Array 1450796_at - -
          Mouse Genome 430 2.0 Array 1450796_PM_at - -
          Mouse Genome 430 2.0 Array 1422929_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53404_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53404_at - -
          Affymetrix Mouse Clariom S Array PSR1000004104.mm.1 NM_016864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004104.mm.1 NM_016864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004104.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004105.mm.1 NM_016864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004105.mm.1 NM_016864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004105.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004106.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004103.mm.1 NM_016864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004103.mm.1 NM_016864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004103.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004108.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004102.mm.1 TC1000000597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004107.mm.1 TC1000000597.mm.1 MTA TC
          Mouse Expression Set 430 1450796_PM_at - -
          Mouse Expression Set 430 1422929_s_at - -
          Mouse Expression Set 430 1450796_at - -
          Mouse Expression Set 430 1422929_PM_s_at - -
          Murine Genome U74v2 Set 101811_at - -
          Murine Genome U74v2 Set 101811_PM_at - -
          Murine Genome U74v2 Set 139808_at - -
          Murine Genome U74v2 Set 139808_PM_at - -
          Murine Genome U74v2 Set 139124_PM_f_at - -
          Murine Genome U74v2 Set 139124_f_at - -
          Mouse Genome 430A 2.0 Array 1450796_at - -
          Mouse Genome 430A 2.0 Array 1422929_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1450796_PM_at - -
          Mouse Genome 430A 2.0 Array 1422929_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450796_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422929_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450796_at - -
          Affymetrix HT Mouse Genome 430A Array 1422929_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 685278 - -
          ABI Mouse Genome Survey Microarray 768248 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550121 - -
          Illumina mouse-6 v1.0 expression beadchip 5550121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247770 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247770 - -
          mouse-6 v1.1 (Illumina) scl38853.1.207_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38853.1.207_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247770 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline