Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850074_g1
          See other GPX2 GE Assays ›
          Gene Symbol:
          Gpx2
          Gene Name:
          glutathione peroxidase 2
          Gene Aliases:
          GI-GPx, GPx-GI, GSHPx-2, GSHPx-GI
          Chromosome Location:
          Chr.12: 76792335 - 76795554 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850074_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030677.2 NP_109602.2 2 886 147
          GenBank mRNA AK131940.1 - 2 763 147
          BC010823.1 - 2 756 4218623 147
          BC034335.1 - 2 733 5038341 147
          BC039658.1 - 2 831 4987478 147
          BC054848.1 - 2 709 6529267 147
          BC066825.1 - 1 2983 147
          BX629737.1 - 1 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpx2

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          negative regulation of inflammatory response to antigenic stimulus
          response to oxidative stress
          response to symbiotic bacterium
          interaction with symbiont
          oxidation-reduction process
          peroxidase activity
          glutathione peroxidase activity
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BC066825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BX629737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 NM_030677.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 AK131940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BC066825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BX629737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 NM_030677.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 AK131940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47354 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101566 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BC066825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BX629737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 NM_030677.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 AK131940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BC010823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BC066825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BX629737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 NM_030677.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 AK131940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BC066825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BX629737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 NM_030677.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 AK131940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BC066825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BX629737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 NM_030677.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 AK131940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28555955 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101566 - -
          Whole Mouse Genome G4122A A_51_P486810 BC066825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 BX629737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 NM_030677.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 AK131940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 BC010823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 BC066825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 BX629737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 NM_030677.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 AK131940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486810 BC010823.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117693 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139484 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09839375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09839361 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09839389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09839382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09839370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28555924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09976706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09840828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10192838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10245553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10192823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10358996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28555936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401113 - -
          Affymetrix HT MG-430 PM Array Plate 1449279_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401109 - -
          Mouse Genome 430 2.0 Array 1449279_at - -
          Mouse Genome 430 2.0 Array 1449279_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14776_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14776_PM_at - -
          Mouse Expression Set 430 1449279_at - -
          Mouse Expression Set 430 1449279_PM_at - -
          Murine Genome U74v2 Set 99810_at - -
          Murine Genome U74v2 Set 99810_PM_at - -
          Mouse Genome 430A 2.0 Array 1449279_PM_at - -
          Mouse Genome 430A 2.0 Array 1449279_at - -
          Affymetrix HT Mouse Genome 430A Array 1449279_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449279_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892390 - -
          ABI Mouse Genome Survey Microarray 903737 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940619 - -
          Illumina mouse-6 v1.0 expression beadchip 6940619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674483 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674483 - -
          mouse-6 v1.1 (Illumina) scl014776.1_184-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014776.1_184-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674483 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014776.1_184-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline